siRNA Details
Gene Symbol: DENND2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318052.1 | NP_001304981.1 | 2 | 699 | ||
NM_001318053.1 | NP_001304982.1 | 3 | 609 | |||
NM_015689.4 | NP_056504.3 | 3 | 801 | |||
NR_134477.1 | 1 | 426 | ||||
XM_005249977.3 | XP_005250034.1 | 3 | 1332 | |||
XM_005249979.4 | XP_005250036.1 | 3 | 585 | |||
XM_011516052.2 | XP_011514354.1 | 3 | 679 | |||
XM_011516053.2 | XP_011514355.1 | 2 | 783 | |||
XM_011516054.2 | XP_011514356.1 | 3 | 587 | |||
XM_017011989.1 | XP_016867478.1 | 2 | 785 | |||
GenBank mRNA | AB033103.2 | - | 2 | 699 | ||
AK290056.1 | - | 3 | 570 | |||
AK297959.1 | - | 2 | 468 | |||
BC115004.1 | - | 3 | 609 | 7470138 | ||
BC132875.1 | - | 1 | 396 | 40146897 | ||
DA401015.1 | - | 1 | 141 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
