siRNA Details
Gene Symbol: DENND2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318052.1 | NP_001304981.1 | 9, 10 | 2307 | ||
NM_001318053.1 | NP_001304982.1 | 10, 11 | 2217 | |||
NM_015689.4 | NP_056504.3 | 10, 11 | 2409 | |||
NR_134477.1 | 8, 9 | 2034 | ||||
XM_005249977.3 | XP_005250034.1 | 10, 11 | 2940 | |||
XM_005249979.4 | XP_005250036.1 | 10, 11 | 2193 | |||
XM_011516052.2 | XP_011514354.1 | 10, 11 | 2287 | |||
XM_011516053.2 | XP_011514355.1 | 9, 10 | 2391 | |||
XM_011516054.2 | XP_011514356.1 | 10, 11 | 2192 | |||
XM_017011989.1 | XP_016867478.1 | 9, 10 | 2393 | |||
GenBank mRNA | AB033103.2 | - | 9, 10 | 2307 | ||
AK290056.1 | - | 10, 11 | 2178 | |||
BC049193.1 | - | 6, 7 | 777 | |||
BC115004.1 | - | 10, 11 | 2217 | 7470138 | ||
BC132875.1 | - | 8, 9 | 2004 | 40146897 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
