siRNA Details
Gene Symbol: ZNF630 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037735.3 | NP_001032824.2 | 5 | 707 | ||
NM_001190255.2 | NP_001177184.1 | 5 | 781 | |||
NM_001282201.1 | NP_001269130.1 | 5 | 1022 | |||
NM_001282202.1 | NP_001269131.1 | 5 | 1018 | |||
GenBank mRNA | AK000580.1 | - | 3 | 438 | ||
AK307979.1 | - | 5 | 682 | |||
BC037316.1 | - | 5 | 792 | |||
BC112139.1 | - | 4 | 520 | 8327607 | ||
BX419623.2 | - | 5 | 755 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
