siRNA Details
Gene Symbol: RAB41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001032726.2 | NP_001027898.2 | 5 | 418 | ||
XM_011530948.2 | XP_011529250.1 | 5 | 1615 | |||
XM_017029488.1 | XP_016884977.1 | 5 | 394 | |||
GenBank mRNA | BC117239.1 | - | 5 | 418 | 40125790 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
