siRNA Details
Gene Symbol: BEX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080425.3 | NP_001073894.1 | 3 | 599 | ||
NM_001127688.2 | NP_001121160.1 | 2 | 516 | |||
GenBank mRNA | AK000959.1 | - | 3 | 554 | ||
AL544124.3 | - | 2 | 492 | |||
AY833563.1 | - | 3 | 554 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
