siRNA Details
Gene Symbol: RIPPLY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138382.2 | NP_612391.1 | 1, 2 | 207 | ||
GenBank mRNA | BC007652.1 | - | 1, 2 | 193 | ||
BC101998.1 | - | 1, 2 | 207 | |||
BC105691.1 | - | 1, 2 | 207 | 40023299 | ||
BC105692.1 | - | 1, 2 | 207 | 40023298 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
