siRNA Details
Gene Symbol: FRMPD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032428.1 | NP_115804.1 | 16 | 4988 | ||
XM_006724709.2 | XP_006724772.1 | 10 | 4463 | |||
XM_011531056.2 | XP_011529358.1 | 14 | 4964 | |||
XM_011531057.2 | XP_011529359.1 | 14 | 4890 | |||
XM_011531058.2 | XP_011529360.1 | 13 | 4802 | |||
XM_017029901.1 | XP_016885390.1 | 14 | 5209 | |||
XM_017029902.1 | XP_016885391.1 | 13 | 5119 | |||
GenBank mRNA | AB058720.1 | - | 1 | 2607 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
