siRNA Details
Gene Symbol: ARHGAP39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308207.1 | NP_001295136.1 | 6 | 2425 | ||
NM_001308208.1 | NP_001295137.1 | 5 | 2078 | |||
NM_025251.2 | NP_079527.1 | 4 | 1951 | |||
XM_011517308.1 | XP_011515610.1 | 5 | 2232 | |||
XM_011517309.1 | XP_011515611.1 | 5 | 2136 | |||
XM_011517312.2 | XP_011515614.1 | 3 | 2770 | |||
XM_017013870.1 | XP_016869359.1 | 6 | 3304 | |||
XM_017013871.1 | XP_016869360.1 | 5 | 2363 | |||
GenBank mRNA | AB051475.1 | - | 6 | 2429 | ||
AK296353.1 | - | 5 | 2075 | |||
AK303851.1 | - | 1 | 654 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
