siRNA Details
Gene Symbol: ARHGAP39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308207.1 | NP_001295136.1 | 12 | 5026 | ||
NM_001308208.1 | NP_001295137.1 | 11 | 4679 | |||
NM_025251.2 | NP_079527.1 | 11 | 4645 | |||
XM_011517308.1 | XP_011515610.1 | 12 | 4926 | |||
XM_011517309.1 | XP_011515611.1 | 12 | 4830 | |||
XM_011517312.2 | XP_011515614.1 | 10 | 5464 | |||
XM_017013870.1 | XP_016869359.1 | 13 | 5998 | |||
XM_017013871.1 | XP_016869360.1 | 12 | 5057 | |||
GenBank mRNA | AB051475.1 | - | 12 | 5030 | ||
AK092501.1 | - | 7 | 2092 | |||
BC013071.2 | - | 5 | 1956 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
