siRNA Details
Gene Symbol: RIC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135920.2 | NP_001129392.2 | 13, 14 | 1686 | ||
NM_020829.3 | NP_065880.2 | 13, 14 | 1686 | |||
XM_005251523.3 | XP_005251580.1 | 13, 14 | 1772 | |||
XM_011517967.2 | XP_011516269.1 | 6, 7 | 684 | |||
XM_017014934.1 | XP_016870423.1 | 13, 14 | 1342 | |||
XM_017014935.1 | XP_016870424.1 | 11, 12 | 2243 | |||
XM_017014936.1 | XP_016870425.1 | 11, 12 | 1182 | |||
GenBank mRNA | AB205401.1 | - | 12, 13 | 1280 | ||
AL136875.1 | - | 13, 14 | 1478 | |||
BC136616.1 | - | 12, 13 | 1280 | 9020606 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
