siRNA Details
Gene Symbol: RIC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135920.2 | NP_001129392.2 | 14 | 1697 | ||
NM_020829.3 | NP_065880.2 | 14 | 1697 | |||
XM_005251523.3 | XP_005251580.1 | 14 | 1783 | |||
XM_011517967.2 | XP_011516269.1 | 7 | 695 | |||
XM_017014934.1 | XP_016870423.1 | 14 | 1353 | |||
XM_017014935.1 | XP_016870424.1 | 12 | 2254 | |||
XM_017014936.1 | XP_016870425.1 | 12 | 1193 | |||
GenBank mRNA | AB205401.1 | - | 13 | 1291 | ||
AL136875.1 | - | 14 | 1489 | |||
BC136616.1 | - | 13 | 1291 | 9020606 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
