siRNA Details
Gene Symbol: PTENP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_023917.1 | 1 | 1165 | |||
GenBank mRNA | AF017999.1 | - | 1 | 396 | ||
AF019083.1 | - | 1 | 396 | |||
BC038293.2 | - | 1 | 649 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
