siRNA Details
Gene Symbol: CEP78 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098802.1 | NP_001092272.1 | 1, 2 | 535 | ||
NM_032171.1 | NP_115547.1 | 1, 2 | 535 | |||
XM_005252263.4 | XP_005252320.1 | 1, 2 | 548 | |||
XM_005252265.2 | XP_005252322.1 | 1, 2 | 548 | |||
XM_017015192.1 | XP_016870681.1 | 1, 2 | 548 | |||
XM_017015193.1 | XP_016870682.1 | 1, 2 | 548 | |||
XM_017015194.1 | XP_016870683.1 | 1, 2 | 548 | |||
XM_017015195.1 | XP_016870684.1 | 1, 2 | 548 | |||
GenBank mRNA | AK022705.1 | - | 1, 2 | 515 | ||
BC058931.1 | - | 1, 2 | 448 | |||
BC128058.1 | - | 1, 2 | 403 | 40078333 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
