siRNA Details
Gene Symbol: SNX30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012994.1 | NP_001013012.1 | 2 | 480 | ||
XM_011518691.2 | XP_011516993.1 | 2 | 517 | |||
XM_017014716.1 | XP_016870205.1 | 2 | 548 | |||
GenBank mRNA | AK127013.1 | - | 1 | 184 | ||
BG699098.1 | - | 2 | 501 | |||
BX367137.2 | - | 2 | 479 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
