siRNA Details
Gene Symbol: SNX30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012994.1 | NP_001013012.1 | 9 | 1724 | ||
GenBank mRNA | AK127013.1 | - | 8 | 1428 | ||
AK128710.1 | - | 4 | 818 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
