siRNA Details
Gene Symbol: HMCN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291815.1 | NP_001278744.1 | 63 | 9652 | ||
XM_011518465.2 | XP_011516767.1 | 63 | 9789 | |||
XM_011518466.2 | XP_011516768.1 | 63 | 9789 | |||
XM_011518467.2 | XP_011516769.1 | 62 | 9612 | |||
XM_011518468.2 | XP_011516770.1 | 63 | 9789 | |||
XM_011518469.2 | XP_011516771.1 | 63 | 9789 | |||
XM_017014585.1 | XP_016870074.1 | 42 | 6458 | |||
XM_017014586.1 | XP_016870075.1 | 17 | 2386 | |||
GenBank mRNA | AL834139.1 | - | 4 | 365 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
