siRNA Details
Gene Symbol: USP6NL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080491.3 | NP_001073960.1 | 7, 8 | 591 | ||
NM_014688.3 | NP_055503.1 | 8, 9 | 922 | |||
XM_006717542.3 | XP_006717605.1 | 9, 10 | 748 | |||
XM_011519762.2 | XP_011518064.1 | 10, 11 | 1081 | |||
XM_011519763.2 | XP_011518065.1 | 3, 4 | 1082 | |||
XM_017016971.1 | XP_016872460.1 | 9, 10 | 755 | |||
XM_017016972.1 | XP_016872461.1 | 9, 10 | 893 | |||
XM_017016973.1 | XP_016872462.1 | 7, 8 | 3918 | |||
GenBank mRNA | AB449916.1 | - | 6, 7 | 492 | ||
AK292944.1 | - | 8, 9 | 712 | |||
BC042943.2 | - | 8, 9 | 760 | 5296060 | ||
CF121539.1 | - | 7, 8 | 598 | |||
D13644.1 | - | 8, 9 | 771 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
