siRNA Details
Gene Symbol: USP6NL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080491.3 | NP_001073960.1 | 14 | 2633 | ||
NM_014688.3 | NP_055503.1 | 15 | 2964 | |||
XM_006717542.3 | XP_006717605.1 | 16 | 2790 | |||
XM_011519762.2 | XP_011518064.1 | 17 | 3123 | |||
XM_011519763.2 | XP_011518065.1 | 10 | 3124 | |||
XM_017016971.1 | XP_016872460.1 | 16 | 2797 | |||
XM_017016972.1 | XP_016872461.1 | 16 | 2935 | |||
XM_017016973.1 | XP_016872462.1 | 14 | 5960 | |||
GenBank mRNA | AK292944.1 | - | 15 | 2754 | ||
BC042943.2 | - | 15 | 2802 | 5296060 | ||
D13644.1 | - | 15 | 2813 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
