siRNA Details
Gene Symbol: PDCD11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014976.1 | NP_055791.1 | 3, 4 | 314 | ||
XM_005269647.3 | XP_005269704.1 | 3, 4 | 354 | |||
XM_011539538.2 | XP_011537840.1 | 3, 4 | 302 | |||
XM_011539539.2 | XP_011537841.1 | 3, 4 | 302 | |||
XM_011539540.1 | XP_011537842.1 | 2, 3 | 168 | |||
GenBank mRNA | AK294971.1 | - | 3, 4 | 343 | ||
AK307418.1 | - | 3, 4 | 327 | |||
BC049838.1 | - | 3, 4 | 315 | |||
BC064486.1 | - | 3, 4 | 307 | |||
BC080560.1 | - | 3, 4 | 307 | |||
BC111040.1 | - | 3, 4 | 269 | |||
D80007.1 | - | 3, 4 | 267 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
