siRNA Details
Gene Symbol: PDCD11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014976.1 | NP_055791.1 | 26 | 3965 | ||
XM_005269647.3 | XP_005269704.1 | 26 | 4005 | |||
XM_011539538.2 | XP_011537840.1 | 26 | 3953 | |||
XM_011539539.2 | XP_011537841.1 | 26 | 3953 | |||
XM_011539540.1 | XP_011537842.1 | 25 | 3819 | |||
GenBank mRNA | BC049838.1 | - | 26 | 3966 | ||
BC064486.1 | - | 26 | 3958 | |||
BC080560.1 | - | 26 | 3958 | |||
BC111040.1 | - | 26 | 3920 | |||
D80007.1 | - | 26 | 3918 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
