siRNA Details
Gene Symbol: ENDOD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015036.2 | NP_055851.1 | 2 | 2645 | ||
GenBank mRNA | AB020637.1 | - | 2 | 2598 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
