siRNA Details
Gene Symbol: TENM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098816.2 | NP_001092286.2 | 32 | 6971 | ||
XM_011544933.2 | XP_011543235.1 | 15 | 4066 | |||
XM_017017525.1 | XP_016873014.1 | 31 | 7242 | |||
XM_017017526.1 | XP_016873015.1 | 29 | 8147 | |||
GenBank mRNA | AB037723.3 | - | 17 | 4249 | ||
AK056531.1 | - | 1 | 632 | |||
AK127101.1 | - | 3 | 1266 | |||
AK127705.1 | - | 5 | 1900 | |||
BX537983.1 | - | 4 | 1415 | |||
BX640737.1 | - | 6 | 2398 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
