siRNA Details
Gene Symbol: ZC3H12C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033390.1 | NP_203748.1 | 4 | 1133 | ||
XM_005271715.3 | XP_005271772.1 | 4 | 1514 | |||
XM_011543055.2 | XP_011541357.1 | 4 | 1246 | |||
XM_017018477.1 | XP_016873966.1 | 4 | 5684 | |||
GenBank mRNA | AB051513.1 | - | 2 | 306 | ||
AB096241.1 | - | 3 | 1870 | |||
AK095101.1 | - | 4 | 1004 | |||
AK295437.1 | - | 4 | 1134 | |||
AK299100.1 | - | 4 | 1166 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
