siRNA Details
Gene Symbol: ZC3H12C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033390.1 | NP_203748.1 | 6 | 3949 | ||
XM_005271715.3 | XP_005271772.1 | 6 | 4330 | |||
XM_011543055.2 | XP_011541357.1 | 6 | 4062 | |||
XM_017018477.1 | XP_016873966.1 | 6 | 8500 | |||
GenBank mRNA | AB051513.1 | - | 4 | 3123 | ||
AB096241.1 | - | 5 | 4686 | |||
AL832893.1 | - | 3 | 2802 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
