siRNA Details
Gene Symbol: C20orf78 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242671.2 | NP_001229600.1 | 2, 3 | 274 | ||
NM_001302818.1 | NP_001289747.1 | 2, 3 | 250 | |||
GenBank mRNA | BX112808.1 | - | 2, 3 | 224 | ||
HY016900.1 | - | 2, 3 | 208 | |||
HY050710.1 | - | 2, 3 | 275 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
