siRNA Details
Gene Symbol: C20orf78 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242671.2 | NP_001229600.1 | 2 | 231 | ||
NM_001302818.1 | NP_001289747.1 | 2 | 207 | |||
GenBank mRNA | BX112808.1 | - | 2 | 181 | ||
BX334499.2 | - | 2 | 192 | |||
CR738909.1 | - | 2 | 223 | |||
HY016900.1 | - | 2 | 165 | |||
HY050710.1 | - | 2 | 232 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
