siRNA Details
Gene Symbol: PLCH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303012.1 | NP_001289941.1 | 22 | 4166 | ||
NM_001303013.1 | NP_001289942.1 | 22 | 4523 | |||
NM_014638.3 | NP_055453.2 | 22 | 3554 | |||
XM_011542455.2 | XP_011540757.1 | 23 | 4097 | |||
XM_011542456.2 | XP_011540758.1 | 23 | 4054 | |||
XM_017002870.1 | XP_016858359.1 | 23 | 4086 | |||
XM_017002871.1 | XP_016858360.1 | 23 | 4057 | |||
XM_017002872.1 | XP_016858361.1 | 23 | 3585 | |||
XM_017002873.1 | XP_016858362.1 | 22 | 3477 | |||
XM_017002874.1 | XP_016858363.1 | 23 | 3562 | |||
GenBank mRNA | AB007919.2 | - | 22 | 4166 | ||
AK074149.1 | - | 10 | 3128 | |||
AK122589.1 | - | 19 | 5165 | |||
AK122591.2 | - | 8 | 3011 | |||
BC019679.1 | - | 8 | 1182 | |||
BC043358.1 | - | 19 | 2748 | |||
BC044630.1 | - | 6 | 935 | |||
BC050037.1 | - | 2 | 536 | |||
BC128207.1 | - | 20 | 2936 | |||
BC128208.1 | - | 6 | 944 | |||
BC144620.1 | - | 22 | 4515 | |||
DQ176850.1 | - | 22 | 3554 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
