siRNA Details
Gene Symbol: TSHZ2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193421.1 | NP_001180350.1 | 3 | 5424 | ||
NM_173485.5 | NP_775756.3 | 3 | 5957 | |||
XM_017027641.1 | XP_016883130.1 | 3 | 5928 | |||
GenBank mRNA | AF230201.1 | - | 1 | 145 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
