siRNA Details
Gene Symbol: KIAA1755 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029864.1 | NP_001025035.1 | 3 | 1338 | ||
GenBank mRNA | AB051542.1 | - | 1 | 796 | ||
AK096202.1 | - | 3 | 1338 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
