siRNA Details
Gene Symbol: OCSTAMP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080721.2 | NP_542452.1 | 3 | 1697 | ||
XM_011528532.2 | XP_011526834.1 | 3 | 1419 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
