siRNA Details
Gene Symbol: SPINT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178455.1 | NP_848550.1 | 1 | 79 | ||
GenBank mRNA | AY372173.1 | - | 1 | 79 | ||
AY372174.1 | - | 1 | 79 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
