siRNA Details
Gene Symbol: TMEM219 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083613.1 | NP_001077082.1 | 4 | 668 | ||
NM_194280.3 | NP_919256.1 | 4 | 665 | |||
XM_005255123.1 | XP_005255180.1 | 4 | 667 | |||
GenBank mRNA | BC017488.1 | - | 3 | 433 | ||
BC031217.2 | - | 3 | 561 | |||
BC050051.1 | - | 4 | 695 | 5278133 | ||
CF453990.1 | - | 612 | ||||
FJ748884.1 | - | 3 | 528 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
