siRNA Details
Gene Symbol: TMEM219 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083613.1 | NP_001077082.1 | 2, 3 | 316 | ||
NM_194280.3 | NP_919256.1 | 2, 3 | 313 | |||
XM_005255123.1 | XP_005255180.1 | 2, 3 | 315 | |||
GenBank mRNA | BC017488.1 | - | 1, 2 | 81 | ||
BC031217.2 | - | 1, 2 | 210 | |||
BC050051.1 | - | 2, 3 | 343 | 5278133 | ||
BF724791.1 | - | 2, 3 | 273 | |||
BU857546.1 | - | 280 | ||||
CF453990.1 | - | 260 | ||||
FJ748884.1 | - | 1, 2 | 176 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
