siRNA Details
Gene Symbol: TMEM219 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083613.1 | NP_001077082.1 | 3 | 485 | ||
NM_194280.3 | NP_919256.1 | 3 | 482 | |||
XM_005255123.1 | XP_005255180.1 | 3 | 484 | |||
GenBank mRNA | BC017488.1 | - | 2 | 250 | ||
BC031217.2 | - | 2 | 379 | |||
BC050051.1 | - | 3 | 512 | 5278133 | ||
BF724791.1 | - | 3 | 442 | |||
CF453990.1 | - | 429 | ||||
FJ748884.1 | - | 2 | 345 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
