siRNA Details
Gene Symbol: KIAA1328 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322327.1 | NP_001309256.1 | 6 | 657 | ||
NM_020776.1 | NP_065827.1 | 7 | 741 | |||
NR_136303.1 | 7 | 752 | ||||
NR_136304.1 | 9 | 1506 | ||||
NR_136305.1 | 6 | 679 | ||||
XM_005258315.4 | XP_005258372.1 | 7 | 744 | |||
XM_005258317.3 | XP_005258374.1 | 7 | 1133 | |||
XM_005258318.4 | XP_005258375.1 | 7 | 748 | |||
XM_011526099.2 | XP_011524401.1 | 7 | 937 | |||
XM_011526100.2 | XP_011524402.1 | 7 | 744 | |||
XM_017025876.1 | XP_016881365.1 | 7 | 1054 | |||
XM_017025877.1 | XP_016881366.1 | 7 | 680 | |||
XM_017025878.1 | XP_016881367.1 | 6 | 834 | |||
XM_017025879.1 | XP_016881368.1 | 6 | 2495 | |||
XM_017025880.1 | XP_016881369.1 | 3 | 2242 | |||
XM_017025881.1 | XP_016881370.1 | 6 | 2325 | |||
GenBank mRNA | AB037749.2 | - | 7 | 1133 | ||
AK292288.1 | - | 7 | 730 | |||
AK304230.1 | - | 6 | 657 | |||
AL832625.2 | - | 7 | 742 | |||
BC008670.1 | - | 1 | 167 | |||
BC037903.1 | - | 3 | 565 | |||
BX648875.1 | - | 6 | 633 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
