siRNA Details
Gene Symbol: KIAA1328 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020776.1 | NP_065827.1 | 3, 4 | 256 | ||
NR_136303.1 | 3, 4 | 267 | ||||
NR_136304.1 | 3, 4 | 267 | ||||
XM_005258315.4 | XP_005258372.1 | 3, 4 | 259 | |||
XM_005258317.3 | XP_005258374.1 | 3, 4 | 648 | |||
XM_005258318.4 | XP_005258375.1 | 3, 4 | 263 | |||
XM_011526099.2 | XP_011524401.1 | 3, 4 | 452 | |||
XM_011526100.2 | XP_011524402.1 | 3, 4 | 259 | |||
XM_017025876.1 | XP_016881365.1 | 3, 4 | 569 | |||
XM_017025877.1 | XP_016881366.1 | 3, 4 | 195 | |||
GenBank mRNA | AB037749.2 | - | 3, 4 | 648 | ||
AK292288.1 | - | 3, 4 | 245 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
