siRNA Details
Gene Symbol: PPP1R1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001261424.1 | NP_001248353.1 | 6 | 916 | ||
NM_001261425.1 | NP_001248354.1 | 6 | 895 | |||
NR_048566.1 | 6 | 1062 | ||||
GenBank mRNA | AF494535.1 | - | 6 | 863 | ||
AK129898.1 | - | 6 | 851 | |||
BC017943.1 | - | 6 | 864 | 4288919 | ||
BX647581.1 | - | 6 | 828 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
