siRNA Details
Gene Symbol: PPP1R1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080545.2 | NP_001074014.1 | 4 | 519 | ||
NM_001261424.1 | NP_001248353.1 | 4 | 540 | |||
NM_001261425.1 | NP_001248354.1 | 4 | 519 | |||
NR_048566.1 | 4 | 519 | ||||
NR_048567.1 | 4 | 530 | ||||
XM_017003451.1 | XP_016858940.1 | 3 | 373 | |||
GenBank mRNA | AF494535.1 | - | 4 | 487 | ||
AK129898.1 | - | 4 | 475 | |||
AK311571.1 | - | 4 | 530 | |||
BC017943.1 | - | 4 | 321 | 4288919 | ||
BX647581.1 | - | 4 | 452 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
