siRNA Details
Gene Symbol: PRSS53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011545817.1 | XP_011544119.1 | 9 | 3135 | ||
XM_011545820.1 | XP_011544122.1 | 10 | 2994 | |||
GenBank mRNA | AK131087.1 | - | 2 | 4936 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
