siRNA Details
Gene Symbol: GXYLT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099650.1 | NP_001093120.1 | 3 | 632 | ||
NM_173601.1 | NP_775872.1 | 4 | 725 | |||
XM_017019211.1 | XP_016874700.1 | 4 | 464 | |||
GenBank mRNA | AK125141.1 | - | 3 | 485 | ||
BC030023.1 | - | 4 | 733 | 5266607 | ||
BC039145.1 | - | 3 | 472 | 4508300 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
