siRNA Details
Gene Symbol: GXYLT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099650.1 | NP_001093120.1 | 7 | 1903 | ||
NM_173601.1 | NP_775872.1 | 8 | 1996 | |||
XM_017019211.1 | XP_016874700.1 | 8 | 1735 | |||
GenBank mRNA | AK125141.1 | - | 7 | 1756 | ||
AL832209.1 | - | 4 | 1133 | |||
BC015597.2 | - | 4 | 1058 | |||
BC030023.1 | - | 8 | 2004 | 5266607 | ||
BC039145.1 | - | 7 | 1743 | 4508300 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
