siRNA Details
Gene Symbol: AGAP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133447.1 | NP_597704.1 | 12 | 1410 | ||
GenBank mRNA | AB075855.1 | - | 7 | 2814 | ||
AK097165.1 | - | 12 | 1410 | |||
BC140762.1 | - | 8 | 656 | 9021630 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
