siRNA Details
Gene Symbol: HEATR5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015473.3 | NP_056288.2 | 9, 10 | 1532 | ||
GenBank mRNA | AK092400.1 | - | 2, 3 | 274 | ||
BC045764.1 | - | 3, 4 | 545 | |||
CR749444.1 | - | 2, 3 | 230 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
