siRNA Details
Gene Symbol: HEATR5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015473.3 | NP_056288.2 | 15 | 2401 | ||
GenBank mRNA | AB037737.1 | - | 6 | 849 | ||
AK092400.1 | - | 8 | 1143 | |||
BC045764.1 | - | 9 | 1414 | |||
CR749444.1 | - | 8 | 1099 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
