siRNA Details
Gene Symbol: HEATR5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015473.3 | NP_056288.2 | 27 | 4383 | ||
GenBank mRNA | AB037737.1 | - | 18 | 2831 | ||
AK000343.1 | - | 5 | 669 | |||
AK023789.1 | - | 9 | 1390 | |||
BC045764.1 | - | 21 | 3396 | |||
BC063867.1 | - | 9 | 1407 | |||
CR749444.1 | - | 20 | 3081 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
