siRNA Details
Gene Symbol: CEP128 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152446.3 | NP_689659.2 | 24 | 4334 | ||
XM_006720056.3 | XP_006720119.1 | 25 | 4515 | |||
XM_011536490.2 | XP_011534792.1 | 26 | 4605 | |||
XM_011536492.2 | XP_011534794.1 | 26 | 4446 | |||
XM_017021042.1 | XP_016876531.1 | 27 | 4723 | |||
XM_017021043.1 | XP_016876532.1 | 14 | 3068 | |||
GenBank mRNA | AK056756.1 | - | 11 | 2025 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
