siRNA Details
Gene Symbol: CEP128 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152446.3 | NP_689659.2 | 22 | 3229 | ||
XM_006720056.3 | XP_006720119.1 | 23 | 3410 | |||
XM_011536490.2 | XP_011534792.1 | 24 | 3500 | |||
XM_011536491.2 | XP_011534793.1 | 24 | 3500 | |||
XM_011536492.2 | XP_011534794.1 | 24 | 3341 | |||
XM_011536493.2 | XP_011534795.1 | 24 | 3500 | |||
XM_017021042.1 | XP_016876531.1 | 25 | 3618 | |||
XM_017021043.1 | XP_016876532.1 | 12 | 1963 | |||
GenBank mRNA | AK056756.1 | - | 9 | 918 | ||
AK125925.1 | - | 13 | 2226 | |||
BC150610.1 | - | 21 | 3087 | 9056980 | ||
HQ215523.1 | - | 24 | 3348 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
