siRNA Details
Gene Symbol: ANKRD50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167882.1 | NP_001161354.1 | 3 | 552 | ||
NM_020337.2 | NP_065070.1 | 4 | 1827 | |||
XM_017008471.1 | XP_016863960.1 | 3 | 2560 | |||
GenBank mRNA | AK127496.1 | - | 3 | 678 | ||
AK291068.1 | - | 4 | 1827 | |||
AK295135.1 | - | 3 | 552 | |||
BX640674.1 | - | 3 | 344 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
