siRNA Details
Gene Symbol: FAM160B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135051.1 | NP_001128523.1 | 3 | 508 | ||
NM_020940.3 | NP_065991.3 | 3 | 508 | |||
GenBank mRNA | AB046820.2 | - | 2 | 103 | ||
BC037207.1 | - | 3 | 508 | 4828347 | ||
CR933692.1 | - | 3 | 228 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
