siRNA Details
Gene Symbol: VWA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009814.1 | NP_001009814.1 | 14 | 1669 | ||
NM_015058.1 | NP_055873.1 | 14 | 1669 | |||
XM_006719791.3 | XP_006719854.1 | 6 | 625 | |||
XM_011535007.2 | XP_011533309.1 | 14 | 1695 | |||
XM_017020469.1 | XP_016875958.1 | 16 | 1728 | |||
XM_017020470.1 | XP_016875959.1 | 14 | 1695 | |||
XM_017020471.1 | XP_016875960.1 | 14 | 1695 | |||
XM_017020474.1 | XP_016875963.1 | 14 | 1695 | |||
GenBank mRNA | AB011136.1 | - | 3 | 209 | ||
AK095155.1 | - | 14 | 1543 | |||
AL834181.1 | - | 7 | 718 | |||
BC053674.1 | - | 14 | 1669 | 6171968 | ||
BC131802.1 | - | 15 | 1502 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
